cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
: TCI
:53292-90-3
:
:C₁₂H₂₁NO₄
:MFCD01862294
:243.30 g/mol
:169°C
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together