
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]triphenylphosphine nickel(II) dichloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:903592-98-3
:
Formula:C₄₅H₅₁Cl₂N₂NiP
MDL Number:MFCD12545958
:780.48 g/mol
:GHS08
Specification Test Results
- Appearance:Yellow to Amber to Dark red powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together