Order Entry
Netherlands
ContactUsLinkComponent
 
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥96.0% (by titrimetric analysis)
 :  TCI
 
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥96.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Chelometric Titration):
    min. 96.0 %

 

Frequently Bought Together