(S)-(-)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:142128-92-5
:
Formula:C₂₄H₂₂O₄
MDL Number:MFCD03788935
:374.44 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:100.0 to 104.0 deg-C
- Specific rotation [a]20/D:-90.0 to -95.0 deg (C=1,CHCl3)
Specification Test Results
Frequently Bought Together