Order Entry
Netherlands
ContactUsLinkComponent
(R,R'')-2,2''-Bis(diphenylphosphino)-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
(R,R'')-2,2''-Bis(diphenylphosphino)-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
(R,R'')-2,2''-Bis(diphenylphosphino)-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -151.0 to -167.0 deg (C=0.2, CHCl3)

Specification Test Results

Related Information

Frequently Bought Together