
N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine ≥90.0% (by HPLC)
TCIAB3174-50MG
: TCI
:
- Pk:50 mg
- Seq:2370
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 90.0 area%
- Elemental analysis (Nitrogen):12.00 to 14.50 %
- Protein labeling test (qualitative methoed):to pass test
:
:525.62 g/mol
:GHS07
:Freezer
:305372-39-8
:C₂₃H₃₅N₅O₇S
:MFCD09264743