
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:95408-45-0
:
Formula:C₂₆H₄₄Cl₂FeP₂Pd
MDL Number:MFCD08064219
:651.75 g/mol
:GHS07
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together