2-(4-Bromophenyl)-1,1-diphenylethylene ≥98.0% (by GC)
Catalog # TCIAB3090-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:77.0 to 81.0 deg-C
Specification Test Results
About this item
:
:335.24 g/mol
CAS Number:18648-66-3
Formula:C₂₀H₁₅Br
MDL Number:MFCD01790865