N-(tert-Butoxycarbonyl)-D-methionine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5241-66-7
:
Formula:C₁₀H₁₉NO₄S
MDL Number:MFCD00038256
:249.33 g/mol
EINECS:226-043-7
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Specific rotation [a]20/D:+19.0 to +25.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together