N-(tert-Butoxycarbonyl)-D-tyrosine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:70642-86-3
:
Formula:C₁₄H₁₉NO₅
MDL Number:MFCD00063030
:281.31 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-1.0 to -4.0 deg (C=2, AcOH)
Specification Test Results
Frequently Bought Together