
Bis(2-aminophenyl)sulphide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2928-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
About this item
:
:216.30 g/mol
:GHS07
CAS Number:5873-51-8
Formula:C₁₂H₁₂N₂S
MDL Number:MFCD03094662