Order Entry
Netherlands
ContactUsLinkComponent
Bis(4-cyanobenzyl)amine hydrochloride ≥98.0% (by titrimetric analysis)
Toxic
Bis(4-cyanobenzyl)amine hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2894-1G
Supplier:  TCI
CAS Number:  
Toxic
Bis(4-cyanobenzyl)amine hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2894-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :283.76 g/mol
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:1802566-49-9
Formula:C₁₆H₁₃N₃