6-(tert-Butoxycarbonylamino)-1-hexanol ≥98.0% (by GC)
Catalog # TCIAB2870-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Melting point:36.0 to 42.0 deg-C
Specification Test Results
About this item
:
:217.31 g/mol
Storage Temperature:Refrigerator
CAS Number:75937-12-1
Formula:C₁₁H₂₃NO₃
MDL Number:MFCD01862954