
Bis[2-(diphenylphosphino)phenyl]ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:166330-10-5
:
Formula:C₃₆H₂₈OP₂
MDL Number:MFCD00233863
:538.57 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:184.0 to 188.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together