
1,9-Bis(acryloyloxy)nonane ≥92.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:107481-28-7
:
Formula:C₁₅H₂₄O₄
MDL Number:MFCD00080592
:268.35 g/mol
Flash Pt:164°C
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:484-500-8
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 92.0 %
Specification Test Results
Frequently Bought Together