Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
(1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane ≥95.0% (by GC)
Catalog # TCIAB2779-100MG
Supplier:  TCI
CAS Number:  
Irritant
(1R,3R)-5-(Bromomethylene)-1,3-bis(tert-butyldimethylsilyloxy)cyclohexane ≥95.0% (by GC)
Catalog # TCIAB2779-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :435.59 g/mol
 :GHS07
CAS Number:387834-41-5
Formula:C₁₉H₃₉BrO₂Si₂