Order Entry
Netherlands
ContactUsLinkComponent
1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane ≥98.0% (by GC)
Irritant
1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane ≥98.0% (by GC)
Catalog # TCIAB2753-25G
Supplier:  TCI
CAS Number:  
Irritant
1,1-Bis(4-hydroxy-3-methylphenyl)cyclohexane ≥98.0% (by GC)
Catalog # TCIAB2753-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    191.0 to 195.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :296.41 g/mol
EINECS:219-110-7
 :GHS07
CAS Number:2362-14-3
Formula:C₂₀H₂₄O₂