
1,1-Bis(3-cyclohexyl-4-hydroxyphenyl)cyclohexane ≥98.0% (by HPLC)
Catalog # TCIAB2752-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:164.0 to 168.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:432.65 g/mol
:GHS07
CAS Number:4221-68-5
Formula:C₃₀H₄₀O₂