Order Entry
Netherlands
ContactUsLinkComponent
4,4'-Bis(2-benzoxazolyl)stilbene ≥97.0% (by HPLC, total nitrogen)
Irritant
4,4'-Bis(2-benzoxazolyl)stilbene ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAB2729-25G
Supplier:  TCI
CAS Number:  
Irritant
4,4'-Bis(2-benzoxazolyl)stilbene ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAB2729-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

About this item

 :
 :414.46 g/mol
Melting Pt:355°C
EINECS:216-245-3
 :GHS07
CAS Number:1533-45-5
Formula:C₂₈H₁₈N₂O₂
MDL Number:MFCD00022846