Order Entry
Netherlands
ContactUsLinkComponent
1,3-Bis(2-hydroxyethylsulphonyl)-2-propanol ≥93.0% (by GC)
1,3-Bis(2-hydroxyethylsulphonyl)-2-propanol ≥93.0% (by GC)
Catalog # TCIAB2699-25G
Supplier:  TCI
CAS Number:  
1,3-Bis(2-hydroxyethylsulphonyl)-2-propanol ≥93.0% (by GC)
Catalog # TCIAB2699-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 93.0 %

Specification Test Results

About this item

 :
 :276.32 g/mol
CAS Number:67006-34-2
Formula:C₇H₁₆O₇S₂