
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:132705-51-2
:
Formula:C₁₂H₂₄BrF₆N₃P₂
MDL Number:MFCD00077412
Melting Pt:139°C
:466.2 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together