4,4'-Bis(5-methyl-2-benzoxazolyl)stilbene ≥96.0% (by HPLC), purified by sublimation
Catalog # TCIAB2676-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 96.0 area%
Specification Test Results
About this item
:
:442.52 g/mol
EINECS:219-260-3
CAS Number:2397-00-4
Formula:C₃₀H₂₂N₂O₂
MDL Number:MFCD00191316