Order Entry
Netherlands
ContactUsLinkComponent
1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene ≥98.0% (by GC)
Irritant
Health
Environment
1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
Health
Environment
1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    136.0 to 140.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together