
9,9-Bis(4-aminophenyl)fluorene ≥99.0% (by HPLC, titration analysis), purified by sublimation
Catalog # TCIAB2654-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
Specification Test Results
About this item
:
:348.45 g/mol
Melting Pt:237°C
:GHS07
CAS Number:15499-84-0
Formula:C₂₅H₂₀N₂
MDL Number:MFCD00039156