
2,6-Bis(chloromethyl)-1,4-dimethoxybenzene
Catalog # TCIAB2649-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
Specification Test Results
About this item
:
:235.10 g/mol
:GHS07
CAS Number:1379163-23-1
Formula:C₁₀H₁₂Cl₂O₂