3',6'-Bis(diethylamino)-2-(4-nitrophenyl)spiro[isoindole-1,9'-xanthene]-3-one ≥98.0% (by total nitrogen basis)
Catalog # TCIAB2628-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:197.0 to 201.0 deg-C
Specification Test Results
About this item
:
:562.67 g/mol
EINECS:249-501-8
CAS Number:29199-09-5
Formula:C₃₄H₃₄N₄O₄