Order Entry
Netherlands
ContactUsLinkComponent
4-(n-Butoxy)benzenesulphonyl chloride ≥98.0% (by GC)
Corrosive
4-(n-Butoxy)benzenesulphonyl chloride ≥98.0% (by GC)
Supplier:  TCI
Certificates
Corrosive
4-(n-Butoxy)benzenesulphonyl chloride ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together