N,N'-Bis[3-(tert-butoxycarbonylamino)propyl]glycinamide ≥93.0% (by titrimetric analysis)
TCIAB2419-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (Nonaqueous Titration):min. 93.0 %
:
:388.51 g/mol
:1822851-77-3
:C₁₈H₃₆N₄O₅