Order Entry
Netherlands
ContactUsLinkComponent
 
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥97.0% (by GC)
 :  TCI
 
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together