

Ethyl-cis-3-bromoacrylate ≥98.0% (by GC, titration analysis)
Catalog # TCIAB2362-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:179.01 g/mol
Boiling Pt:67°C
Storage Temperature:Refrigerator
UN:3272
ADR:3,III
:GHS02
:GHS07
CAS Number:31930-34-4
Formula:C₅H₇BrO₂
MDL Number:MFCD00674501