
N-Benzoylmeroquinene-tert-butyl ester ≥98.0% (by total nitrogen basis)
Catalog # TCIAB2361-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:62.0 to 67.0 deg-C
- Specific rotation [a]20/D:+44.0 to +48.0 deg (C=1, MeOH)
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:329.44 g/mol
:GHS07
CAS Number:52346-13-1
Formula:C₂₀H₂₇NO₃