4,4'-Biphenyldithiol ≥98.0% (by HPLC)
TCIAB2285-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:181.0 to 185.0 deg-C
:
:218.33 g/mol
:230-136-8
:6954-27-4
:C₁₂H₁₀S₂
:MFCD00046826