Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-tert-Butoxy-4-chlorobenzene
1-tert-Butoxy-4-chlorobenzene ≥95.0% (by GC)
1-tert-Butoxy-4-chlorobenzene ≥95.0% (by GC)
TCIAB2278-25G
:
TCI
:
1-tert-Butoxy-4-chlorobenzene ≥95.0% (by GC)
TCIAB2278-25G
:
TCI
:
Pk:
25 g
Seq:
3308
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 95.0 %
:
:
184.66 g/mol
:
18995-35-2
:
C₁₀H₁₃ClO
:
MFCD04038411