Order Entry
Netherlands
ContactUsLinkComponent
4,5-Bis(2-cyanoethylthio)-1,3-dithiol-2-one ≥98.0% (by HPLC, total nitrogen)
Toxic
4,5-Bis(2-cyanoethylthio)-1,3-dithiol-2-one ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB2233-1G
Supplier:  TCI
CAS Number:  
Toxic
4,5-Bis(2-cyanoethylthio)-1,3-dithiol-2-one ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB2233-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    84.0 to 88.0 deg-C

Specification Test Results

About this item

 :
 :288.42 g/mol
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:158871-28-4
Formula:C₉H₈N₂OS₄
MDL Number:MFCD01142888