
N-(tert-Butoxycarbonyl)aminoferrocene ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2227-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
About this item
:
:301.17 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:339195-19-6
Formula:C₁₅H₁₉FeNO₂
MDL Number:MFCD03844774