Order Entry
Netherlands
ContactUsLinkComponent
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥96.0% (by titrimetric analysis)
Irritant
Health
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥96.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
Health
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥96.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (Chelometric Titration):
    min. 96.0 %

Specification Test Results

Frequently Bought Together