5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate ≥94.0% (by titrimetric analysis)
TCIAB2179-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 94.0 %
- Specific rotation [a]20/D:-13.0 to -19.0 deg (C=2, DMF)
:
:329.39 g/mol
:73821-97-3
:C₁₆H₂₇NO₆
:MFCD00065570