Order Entry
Netherlands
ContactUsLinkComponent
Monosodium bromoisocyanurate hydrate ≥95.0% (by titrimetric analysis)
Corrosive
Monosodium bromoisocyanurate hydrate ≥95.0% (by titrimetric analysis)
Catalog # TCIAB2148-25G
Supplier:  TCI
CAS Number:  
Corrosive
Monosodium bromoisocyanurate hydrate ≥95.0% (by titrimetric analysis)
Catalog # TCIAB2148-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 95.0 % (calcd.on ann. substance)
  • Water (Vaporization methoed):
    max. 13.0 %

Specification Test Results

About this item

 :
 :229.95 g/mol
 :GHS05
CAS Number:164918-61-0
Formula:C₃H₂BrN₃O₃
MDL Number:MFCD04037229