2,2-Bis(3-nitrophenyl)hexafluoropropane ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:64465-34-5
:
Formula:C₁₅H₈F₆N₂O₄
MDL Number:MFCD02093448
:394.23 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
Specification Test Results
Frequently Bought Together