1,3-Bis(benzyloxy)-2-propanol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6972-79-8
:
Formula:C₁₇H₂₀O₃
MDL Number:MFCD00010616
:272.34 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together