2-(4-Bromophenyl)-1,3-dioxolane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10602-01-4
:
Formula:C₉H₉BrO₂
MDL Number:MFCD01318950
:229.07 g/mol
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together