Order Entry
Netherlands
ContactUsLinkComponent
1,4-Bis[4-(di-p-tolylamino)styryl]benzene ≥98.0% (by HPLC)
1,4-Bis[4-(di-p-tolylamino)styryl]benzene ≥98.0% (by HPLC)
Catalog # TCIAB2080-1G
Supplier:  TCI
CAS Number:  
1,4-Bis[4-(di-p-tolylamino)styryl]benzene ≥98.0% (by HPLC)
Catalog # TCIAB2080-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    220.0 to 226.0 deg-C

Specification Test Results

About this item

 :
 :672.92 g/mol
EINECS:259-446-1
CAS Number:55035-43-3
Formula:C₅₀H₄₄N₂
MDL Number:MFCD03844769