Order Entry
Netherlands
ContactUsLinkComponent
Biphenyl-2,2'-dicarboxaldehyde ≥97.0% (by GC)
Corrosive
Biphenyl-2,2'-dicarboxaldehyde ≥97.0% (by GC)
Catalog # TCIAB2056-100MG
Supplier:  TCI
CAS Number:  
Corrosive
Biphenyl-2,2'-dicarboxaldehyde ≥97.0% (by GC)
Catalog # TCIAB2056-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    62.0 to 65.0 deg-C

Specification Test Results

About this item

 :
 :210.23 g/mol
Boiling Pt:179°C
 :GHS05
CAS Number:1210-05-5
Formula:C₁₄H₁₀O₂
MDL Number:MFCD00142732