Order Entry
Netherlands
ContactUsLinkComponent
(S,S)-(-)-Bis(ɑ-methylbenzyl)amine hydrochloride ≥98.0% (by HPLC, titration analysis)
Irritant
(S,S)-(-)-Bis(ɑ-methylbenzyl)amine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2035-5G
Supplier:  TCI
CAS Number:  
Irritant
(S,S)-(-)-Bis(ɑ-methylbenzyl)amine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2035-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -70.0 to -76.0 deg (C=3, EtOH)

Specification Test Results

About this item

 :
 :261.79 g/mol
 :GHS07
CAS Number:40648-92-8
Formula:C₁₆H₁₉N
MDL Number:MFCD00216671