Order Entry
Netherlands
ContactUsLinkComponent
 
 
(R,R)-(+)-Bis(ɑ-methylbenzyl)amine hydrochloride ≥98.0% (by HPLC, titration analysis)
  TCIAB2034-5G
 :  TCI
 :  
 
(R,R)-(+)-Bis(ɑ-methylbenzyl)amine hydrochloride ≥98.0% (by HPLC, titration analysis)
  TCIAB2034-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +70.0 to +76.0 deg (C=3, EtOH)

 

 

 :
 :261.79 g/mol
 :GHS07
 :82398-30-9
 :C₁₆H₁₉N
 :MFCD00216672