sec-Butyl methyl ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6795-87-5
:
Formula:C₅H₁₂O
MDL Number:MFCD00190205
Boiling Pt:59°C
Molecular Weight:88.15 g/mol
Flash Pt:–30°C
UN:3271
ADR:3 II
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together