Order Entry
Netherlands
ContactUsLinkComponent
2,6-Bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol ≥98.0% (by titrimetric analysis)
Irritant
2,6-Bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1972-25G
Supplier:  TCI
CAS Number:  
Irritant
2,6-Bis[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1972-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    211.0 to 217.0 deg-C
  • Solubility in N,N-DMF:
    almost transparency

Specification Test Results

About this item

 :
 :348.44 g/mol
EINECS:216-587-3
 :GHS07
CAS Number:1620-68-4
Formula:C₂₃H₂₄O₃
MDL Number:MFCD00155175