
Bis(2,6-dimethylphenyl)chlorophosphate ≥93.0% (by titrimetric analysis)
Catalog # TCIAB1922-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (Argentmetric Titration):min. 93.0 %
- Melting point:50.0 to 54.0 deg-C
Specification Test Results
About this item
:
:324.74 g/mol
Boiling Pt:219°C
UN:1759
ADR:8,III
:GHS05
CAS Number:81639-99-8
Formula:C₁₆H₁₈ClO₃P
MDL Number:MFCD00623313