
N,O-Bis(tert-butyldimethylsilyl)acetamide ≥96.0% (by GC) tert-butyl-dimethyl-silylating agent
Supplier: TCI
Certificates
About this item
CAS Number:82112-21-8
:
Formula:C₁₄H₃₃NOSi₂
MDL Number:MFCD01321138
:287.59 g/mol
UN:1760
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Related Information
Frequently Bought Together