



sec-Butylmagnesium bromide ∼16% ∼1 mol/L in THF
Catalog # TCIAB1884-100G
Supplier: TCI
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Concentration (sec Butanol method):15.0 to 18.0 %
Specification Test Results
About this item
:
:161.33 g/mol
EINECS:213-081-4
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:922-66-7
Formula:C₄H₉BrMg
MDL Number:MFCD01321135